Product Details
- SNP ID
-
rs113215414
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:29724315 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCGCTCCTGGACCGCGGCGGACAC[C/G]GTGGCTCAGATCACCCAGCGCTTCT
- Phenotype
-
MIM: 143110
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HLA-F
PubMed Links
Gene Details
- Gene
- HLA-F
- Gene Name
- major histocompatibility complex, class I, F
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098478.1 |
507 |
Silent Mutation |
ACC,ACG |
T159T |
NP_001091948.1 |
NM_001098479.1 |
507 |
Silent Mutation |
ACC,ACG |
T159T |
NP_001091949.1 |
NM_018950.2 |
507 |
Silent Mutation |
ACC,ACG |
T159T |
NP_061823.2 |
XM_011514564.1 |
507 |
Silent Mutation |
ACC,ACG |
T159T |
XP_011512866.1 |
XM_017010810.1 |
507 |
Silent Mutation |
ACC,ACG |
T159T |
XP_016866299.1 |
XM_017010811.1 |
507 |
Silent Mutation |
ACC,ACG |
T159T |
XP_016866300.1 |
XM_017010812.1 |
507 |
Silent Mutation |
ACC,ACG |
T159T |
XP_016866301.1 |
XM_017010813.1 |
507 |
Silent Mutation |
ACC,ACG |
T159T |
XP_016866302.1 |
XM_017010814.1 |
507 |
Silent Mutation |
ACC,ACG |
T41T |
XP_016866303.1 |
XM_017010815.1 |
507 |
Silent Mutation |
ACC,ACG |
T41T |
XP_016866304.1 |
- Gene
- HLA-F-AS1
- Gene Name
- HLA-F antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details