Product Details

SNP ID
rs115995016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33292132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGCCCGTGCAATCTTCCTCCC[C/T]GTGGCCTTGATCCTGGGAAAGGAGC
Phenotype
MIM: 605660 MIM: 602306 MIM: 601962 MIM: 611440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6834 PubMed Links

Gene Details

Gene
MIR6834
Gene Name
microRNA 6834
There are no transcripts associated with this gene.

Gene
MIR6873
Gene Name
microRNA 6873
There are no transcripts associated with this gene.

Gene
PFDN6
Gene Name
prefoldin subunit 6
There are no transcripts associated with this gene.

Gene
RGL2
Gene Name
ral guanine nucleotide dissociation stimulator like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243738.1 2667 Silent Mutation ACA,ACG T686T NP_001230667.1
NM_004761.4 2667 Silent Mutation ACA,ACG T768T NP_004752.1
XM_011514781.1 2667 Silent Mutation ACA,ACG T661T XP_011513083.1
XM_011514785.2 2667 Intron XP_011513087.1
Gene
TAPBP
Gene Name
TAP binding protein (tapasin)
There are no transcripts associated with this gene.

Gene
WDR46
Gene Name
WD repeat domain 46
There are no transcripts associated with this gene.

View Full Product Details