Product Details

SNP ID
rs114700977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34901016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTCATCTCATGTAGGTGCACGT[A/G]TTCATGAATTTTACCTGTGAGAAAA
Phenotype
MIM: 608994
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKS1A PubMed Links

Gene Details

Gene
ANKS1A
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015245.2 Intron NP_056060.2
XM_005248964.3 Intron XP_005249021.1
XM_006715036.2 Intron XP_006715099.1
XM_011514431.2 Intron XP_011512733.1
XM_011514432.2 Intron XP_011512734.1
XM_011514433.1 Intron XP_011512735.1
XM_011514434.2 Intron XP_011512736.1
XM_011514435.2 Intron XP_011512737.1
XM_011514436.2 Intron XP_011512738.1
XM_011514437.2 Intron XP_011512739.1
XM_017010593.1 Intron XP_016866082.1

View Full Product Details