Product Details

SNP ID
rs116676108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35474380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTGCGGCCAGCCCCCGCCCCAG[A/G]GAGGTAGTATAAACCTGGGCTCTGG
Phenotype
MIM: 613976 MIM: 615660 MIM: 603170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FANCE PubMed Links

Gene Details

Gene
FANCE
Gene Name
Fanconi anemia complementation group E
There are no transcripts associated with this gene.

Gene
MIR7111
Gene Name
microRNA 7111
There are no transcripts associated with this gene.

Gene
RPL10A
Gene Name
ribosomal protein L10a
There are no transcripts associated with this gene.

Gene
TEAD3
Gene Name
TEA domain transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003214.3 2208 UTR 3 NP_003205.2

View Full Product Details