Product Details

SNP ID
rs114035152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36037399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTTGGGCATGCTTGGCTCCAAG[A/G]CTGAATGTTGGGGGAAAGAAGTATA
Phenotype
MIM: 600289
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPK14 PubMed Links

Gene Details

Gene
MAPK14
Gene Name
mitogen-activated protein kinase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001315.2 Intron NP_001306.1
NM_139012.2 Intron NP_620581.1
NM_139013.2 Intron NP_620582.1
NM_139014.2 Intron NP_620583.1
XM_006714998.3 Intron XP_006715061.1
XM_011514310.2 Intron XP_011512612.2
XM_017010299.1 Intron XP_016865788.1
XM_017010300.1 Intron XP_016865789.1
XM_017010301.1 Intron XP_016865790.1
XM_017010302.1 Intron XP_016865791.1
XM_017010303.1 Intron XP_016865792.1
XM_017010304.1 Intron XP_016865793.1

View Full Product Details