Product Details

SNP ID
rs114633981
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39105771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATGTGCTGCCCTGGGGCTGAGC[C/T]GCTTCCTAGGATACACAAACAAACA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SAYSD1 PubMed Links

Gene Details

Gene
SAYSD1
Gene Name
SAYSVFN motif domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304793.1 310 Silent Mutation GCA,GCG A4A NP_001291722.1
NM_018322.2 310 Silent Mutation GCA,GCG A71A NP_060792.1

View Full Product Details