Product Details

SNP ID
rs114731895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41068872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAGAGGTCAGTGACTCCATTCT[C/T]CAGACAATGAGACTTCATTGCCTCT
Phenotype
MIM: 604797 MIM: 189903 MIM: 614393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOBEC2 PubMed Links

Gene Details

Gene
APOBEC2
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 2
There are no transcripts associated with this gene.

Gene
NFYA
Gene Name
nuclear transcription factor Y subunit alpha
There are no transcripts associated with this gene.

Gene
OARD1
Gene Name
O-acyl-ADP-ribose deacylase 1
There are no transcripts associated with this gene.

View Full Product Details