Product Details

SNP ID
rs111464322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41770948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGTCAGTGGCACAGGAAAGCTGC[C/T]GTGGCTGCGGATGGCGGCCCGGGTG
Phenotype
MIM: 607744 MIM: 611389
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRS3 PubMed Links

Gene Details

Gene
FRS3
Gene Name
fibroblast growth factor receptor substrate 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006653.4 2260 Missense Mutation AGC,GGC S384G NP_006644.1
XM_011514254.2 2260 Missense Mutation AGC,GGC S384G XP_011512556.1
XM_011514255.2 2260 Missense Mutation AGC,GGC S384G XP_011512557.1
XM_017010193.1 2260 Missense Mutation AGC,GGC S347G XP_016865682.1
Gene
PRICKLE4
Gene Name
prickle planar cell polarity protein 4
There are no transcripts associated with this gene.

View Full Product Details