Product Details

SNP ID
rs111999753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44392764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAAGTTGATGCCAACTCAGTGG[A/G]GGACCATTGCTCCAATCATTGGAAG
Phenotype
MIM: 602868
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC5L PubMed Links

Gene Details

Gene
CDC5L
Gene Name
cell division cycle 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253.3 557 Silent Mutation AGG,GGG R83G NP_001244.1

View Full Product Details