Product Details

SNP ID
rs114426615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:47234782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGAAGCCCTTGTTCTTGTCCTG[C/T]GGGGAAGGCTCCACCGTCAGGAGAG
Phenotype
MIM: 605732
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF21 PubMed Links

Gene Details

Gene
TNFRSF21
Gene Name
TNF receptor superfamily member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014452.4 2062 Silent Mutation CCA,CCG P542P NP_055267.1
XM_017010744.1 2062 Intron XP_016866233.1

View Full Product Details