Product Details

SNP ID
rs113239409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52757864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGACCTAACTCAGAACCTACCTC[C/T]ACTCCAGCTGCAGCCAGGAGCCACC
Phenotype
MIM: 138360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTA2 PubMed Links

Gene Details

Gene
GSTA2
Gene Name
glutathione S-transferase alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000846.4 234 Silent Mutation GTA,GTG V28V NP_000837.3
XM_011514532.2 234 Intron XP_011512834.1

View Full Product Details