Product Details

SNP ID
rs111660749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:5999036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACACCAGGAGCACCGGGAACGC[C/G]GGGAGCAGGGACCCCGCCGCCCCGT
Phenotype
MIM: 607409
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NRN1 PubMed Links

Gene Details

Gene
NRN1
Gene Name
neuritin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278710.1 1411 Silent Mutation CCC,CCG P123P NP_001265639.1
NM_001278711.1 1411 Silent Mutation CCC,CCG P149P NP_001265640.1
NM_016588.2 1411 Silent Mutation CCC,CCG P123P NP_057672.1
XM_006715106.3 1411 Silent Mutation CCC,CCG P184P XP_006715169.1
XM_006715107.3 1411 Silent Mutation CCC,CCG P114P XP_006715170.1

View Full Product Details