Product Details
- SNP ID
-
rs114626881
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:7393262 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACTGGGATGAAGGAGTTGGAAAA[C/G]TCGCCAAGGGTTATGTCTGGAATGG
- Phenotype
-
MIM: 608304
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CAGE1
PubMed Links
Gene Details
- Gene
- CAGE1
- Gene Name
- cancer antigen 1
There are no transcripts associated with this gene.
- Gene
- RIOK1
- Gene Name
- RIO kinase 1
View Full Product Details