Product Details

SNP ID
rs114151270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:158532723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAACAACAAAAACCAGCAACAGG[C/T]GCTTCCTCCAGACACACCACGGCAC
Phenotype
MIM: 601698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR595 PubMed Links

Gene Details

Gene
MIR595
Gene Name
microRNA 595
There are no transcripts associated with this gene.

Gene
PTPRN2
Gene Name
protein tyrosine phosphatase, receptor type N2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308267.1 Intron NP_001295196.1
NM_001308268.1 Intron NP_001295197.1
NM_002847.4 Intron NP_002838.2
NM_130842.3 Intron NP_570857.2
NM_130843.3 Intron NP_570858.2
XM_011516446.1 Intron XP_011514748.1
XM_011516447.2 Intron XP_011514749.1
XM_011516448.2 Intron XP_011514750.1
XM_011516449.2 Intron XP_011514751.1
XM_017012475.1 Intron XP_016867964.1
XM_017012476.1 Intron XP_016867965.1

View Full Product Details