Product Details

SNP ID
rs116134185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:158733095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGCTGAGAACAGCAGGGGCCG[A/G]GCTCGCAGCTGTGGCGCCCCCAACC
Phenotype
MIM: 616691
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESYT2 PubMed Links

Gene Details

Gene
ESYT2
Gene Name
extended synaptotagmin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020728.2 3860 UTR 3 NP_065779.1

View Full Product Details