Product Details

SNP ID
rs118131793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:11373677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAAATATTTTCATTTTGGGGAC[A/T]ATGACATCCTAACCTGATGTCTTTT
Phenotype
MIM: 612249
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
THSD7A PubMed Links

Gene Details

Gene
THSD7A
Gene Name
thrombospondin type 1 domain containing 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015204.2 7108 UTR 3 NP_056019.1
XM_006715659.1 7108 UTR 3 XP_006715722.1
XM_006715660.1 7108 UTR 3 XP_006715723.1
XM_006715662.1 7108 UTR 3 XP_006715725.1
XM_011515193.2 7108 UTR 3 XP_011513495.1
XM_011515194.1 7108 UTR 3 XP_011513496.1

View Full Product Details