Product Details

SNP ID
rs117847862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1543163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGGGTCAGGATCCGGGCACTAC[A/G]TGTAAGTGGCTTTCCCGGACTGCTG
Phenotype
MIM: 600197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100128653 PubMed Links

Gene Details

Gene
LOC100128653
Gene Name
uncharacterized LOC100128653
There are no transcripts associated with this gene.

Gene
MAFK
Gene Name
MAF bZIP transcription factor K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002360.3 5348 Intron NP_002351.1
XM_005249851.2 5348 Intron XP_005249908.2
XM_006715773.2 5348 Intron XP_006715836.1
Gene
TMEM184A
Gene Name
transmembrane protein 184A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097620.1 5348 UTR 3 NP_001091089.1
XM_006715653.1 5348 Intron XP_006715716.1
XM_006715655.2 5348 Intron XP_006715718.1
XM_011515164.1 5348 Intron XP_011513466.1
XM_011515165.2 5348 Intron XP_011513467.1
XM_011515166.2 5348 Intron XP_011513468.1
XM_017011815.1 5348 Intron XP_016867304.1

View Full Product Details