Product Details

SNP ID
rs114194936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1842485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGGTTCTCCCAGTCCACAGGCC[A/G]CCAGAATCCCTTCCGGCGCATGCCT
Phenotype
MIM: 602686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100127955 PubMed Links

Gene Details

Gene
LOC100127955
Gene Name
uncharacterized LOC100127955
There are no transcripts associated with this gene.

Gene
MAD1L1
Gene Name
MAD1 mitotic arrest deficient like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013836.1 Intron NP_001013858.1
NM_001013837.1 Intron NP_001013859.1
NM_001304523.1 Intron NP_001291452.1
NM_001304524.1 Intron NP_001291453.1
NM_001304525.1 Intron NP_001291454.1
NM_003550.2 Intron NP_003541.2
XM_005249877.1 Intron XP_005249934.1
XM_011515567.1 Intron XP_011513869.1
XM_011515568.2 Intron XP_011513870.1
XM_011515569.2 Intron XP_011513871.2
XM_011515571.1 Intron XP_011513873.1
XM_017012690.1 Intron XP_016868179.1
XM_017012691.1 Intron XP_016868180.1
Gene
MIR4655
Gene Name
microRNA 4655
There are no transcripts associated with this gene.

View Full Product Details