Product Details

SNP ID
rs116780283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:26636463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCATGCACACATATATACCTAG[C/T]CCTCTGTTTTGAGGGCTTAAAAGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf71 PubMed Links

Gene Details

Gene
C7orf71
Gene Name
chromosome 7 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145531.2 882 Intron NP_001139003.1
NM_001302625.1 882 Intron NP_001289554.1
XM_017012010.1 882 UTR 5 XP_016867499.1
XM_017012011.1 882 UTR 5 XP_016867500.1
XM_017012012.1 882 UTR 5 XP_016867501.1
XM_017012013.1 882 UTR 5 XP_016867502.1
XM_017012014.1 882 UTR 5 XP_016867503.1

View Full Product Details