Product Details

SNP ID
rs114286282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2692024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTATGGTGCTTGCCCCTTGTGG[G/T]CGGCAGTGCTGGTGAGTGGAAGGTC
Phenotype
MIM: 615168
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AMZ1 PubMed Links

Gene Details

Gene
AMZ1
Gene Name
archaelysin family metallopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284355.2 Intron NP_001271284.1
NM_001321766.1 Intron NP_001308695.1
NM_133463.3 Intron NP_597720.1
XM_011515150.2 Intron XP_011513452.1
XM_011515151.2 Intron XP_011513453.1
XM_017011774.1 Intron XP_016867263.1
XM_017011775.1 Intron XP_016867264.1
XM_017011776.1 Intron XP_016867265.1

View Full Product Details