Product Details

SNP ID
rs117428786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2906704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGCCGATCTTGTCCTTGACAAC[A/G]CGGAGCAGCTCCTCTACGCTGCCCC
Phenotype
MIM: 607210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARD11 PubMed Links

Gene Details

Gene
CARD11
Gene Name
caspase recruitment domain family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324281.1 3803 Silent Mutation CGC,CGT R1133R NP_001311210.1
NM_032415.5 3803 Silent Mutation CGC,CGT R1133R NP_115791.3
XM_011515586.2 3803 Silent Mutation CGC,CGT R1133R XP_011513888.1
XM_011515587.2 3803 Silent Mutation CGC,CGT R1132R XP_011513889.1

View Full Product Details