Product Details

SNP ID
rs113474959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30753858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCGACCGCAACCGGGAGGAGCT[A/G]GAAAAGTGGCTGAAGAAGGAGCCGG
Phenotype
MIM: 604854
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INMT PubMed Links

Gene Details

Gene
INMT
Gene Name
indolethylamine N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199219.1 295 Silent Mutation CTA,CTG L93L NP_001186148.1
NM_006774.4 295 Silent Mutation CTA,CTG L94L NP_006765.4
Gene
INMT-FAM188B
Gene Name
INMT-FAM188B readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details