Product Details

SNP ID
rs112240167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30912140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCAGCGGCGCCCACCTCAACCC[A/G]GCTGTCACACTGGGGCTGCTGCTCA
Phenotype
MIM: 107776
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AQP1 PubMed Links

Gene Details

Gene
AQP1
Gene Name
aquaporin 1 (Colton blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185060.1 Intron NP_001171989.1
NM_001185061.1 Intron NP_001171990.1
NM_001185062.1 Intron NP_001171991.1

View Full Product Details