Product Details

SNP ID
rs115454435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:32868172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCATAACAAAAGAAAAATAGATA[C/T]TGAATATATACAATTTATTTAATAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KBTBD2 PubMed Links

Gene Details

Gene
KBTBD2
Gene Name
kelch repeat and BTB domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015483.2 3704 UTR 3 NP_056298.2
XM_005249696.2 3704 UTR 3 XP_005249753.1
XM_005249697.2 3704 Intron XP_005249754.1
XM_011515256.2 3704 UTR 3 XP_011513558.1
XM_011515257.2 3704 Intron XP_011513559.1

View Full Product Details