Product Details
- SNP ID
-
rs104894037
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:33096550 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGATGTCATACATGGGATCTTCA[T/A]GTGCCTTAAGGGTCAGAGAAGGTTA
- Phenotype
-
MIM: 607331
- Polymorphism
- T/A, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RP9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17169625] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RP9
- Gene Name
- retinitis pigmentosa 9 (autosomal dominant)
View Full Product Details