Product Details

SNP ID
rs116597131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:38275012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTCTAAATATCACATAAGGGT[C/T]CAGACCAGGCACTTTGGAGTAGACA
Phenotype
MIM: 609642
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TARP PubMed Links

Gene Details

Gene
TARP
Gene Name
TCR gamma alternate reading frame protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003799.1 Intron NP_001003799.1
NM_001003806.1 Intron NP_001003806.1
XM_011515407.2 Intron XP_011513709.1
XM_017012226.1 Intron XP_016867715.1
XM_017012227.1 Intron XP_016867716.1
XM_017012228.1 Intron XP_016867717.1
XM_017012229.1 Intron XP_016867718.1
XM_017012230.1 Intron XP_016867719.1

View Full Product Details