Product Details

SNP ID
rs112599733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44962794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCGAGCGCAGCGGAAATCGGGC[C/T]CTGGCTGCTCCGGCCTGGGCTCCAC
Phenotype
MIM: 600642
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO1G PubMed Links

Gene Details

Gene
MYO1G
Gene Name
myosin IG
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033054.2 2905 Missense Mutation GAG,GGG E1001G NP_149043.2
XM_017012503.1 2905 Missense Mutation GAG,GGG E886G XP_016867992.1
XM_017012504.1 2905 Missense Mutation GAG,GGG E763G XP_016867993.1

View Full Product Details