Product Details

SNP ID
rs115058579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:729774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAAGCCACAGGCGCAGTGATCC[A/T]TTTTGGCAACGGGAAGTCCGTGGAC
Phenotype
MIM: 614864 MIM: 176911
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAAF5 PubMed Links

Gene Details

Gene
DNAAF5
Gene Name
dynein axonemal assembly factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017802.3 727 Missense Mutation CAT,CTT H236L NP_060272.3
Gene
PRKAR1B
Gene Name
protein kinase cAMP-dependent type I regulatory subunit beta
There are no transcripts associated with this gene.

View Full Product Details