Product Details

SNP ID
rs116457847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150330809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCTGTGCGTGGACCCCACCAG[A/G]GTAGCCAAGAGCTCCGGGGTGGAGG
Phenotype
MIM: 601973 MIM: 615252
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTR3C PubMed Links

Gene Details

Gene
ACTR3C
Gene Name
ARP3 actin-related protein 3 homolog C
There are no transcripts associated with this gene.

Gene
LRRC61
Gene Name
leucine rich repeat containing 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142928.1 961 Intron NP_001136400.1
NM_023942.2 961 Intron NP_076431.1
XM_006716095.2 961 Intron XP_006716158.1
Gene
RARRES2
Gene Name
retinoic acid receptor responder 2
There are no transcripts associated with this gene.

Gene
ZBED6CL
Gene Name
ZBED6 C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138434.2 961 Silent Mutation AGA,AGG R135R NP_612443.1

View Full Product Details