Product Details
- SNP ID
-
rs117702478
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:151430104 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGTTGCTGGTTTTGTGGTGGCCT[C/T]TTCCGGTCCTTGATCTGATTGAAGA
- Phenotype
-
MIM: 609603
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRYGN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73476444] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRYGN
- Gene Name
- crystallin gamma N
- Gene
- MIR3907
- Gene Name
- microRNA 3907
There are no transcripts associated with this gene.
View Full Product Details