Product Details

SNP ID
rs115324647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143265779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAAACATTGACCAGTGAGCCAT[A/G]GATGGGGGTATAAAAGTAAGTTTGT
Phenotype
MIM: 602321
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTK1 PubMed Links

Gene Details

Gene
GSTK1
Gene Name
glutathione S-transferase kappa 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143679.1 Intron NP_001137151.1
NM_001143680.1 Intron NP_001137152.1
NM_001143681.1 Intron NP_001137153.1
NM_015917.2 Intron NP_057001.1
Gene
LOC105375545
Gene Name
uncharacterized LOC105375545
There are no transcripts associated with this gene.

View Full Product Details