Product Details

SNP ID
rs112977067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143381651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTACGCCCCGCAGAAGAAGTTCG[A/G]CCCTGTGGTGGCCCCAAAGCCCAAA
Phenotype
MIM: 179610 MIM: 602002
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHA1 PubMed Links

Gene Details

Gene
EPHA1
Gene Name
EPH receptor A1
There are no transcripts associated with this gene.

Gene
FAM131B
Gene Name
family with sequence similarity 131 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031690.2 226 Intron NP_001026860.2
NM_001278297.1 226 Intron NP_001265226.1
NM_014690.4 226 Intron NP_055505.3
XM_005250073.3 226 Intron XP_005250130.1
XM_006716186.2 226 Intron XP_006716249.1
Gene
LOC100507507
Gene Name
uncharacterized LOC100507507
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322263.1 226 Intron NP_001309192.1
Gene
MIR6892
Gene Name
microRNA 6892
There are no transcripts associated with this gene.

Gene
ZYX
Gene Name
zyxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010972.1 226 Missense Mutation GAC,GGC D27G NP_001010972.1
NM_003461.4 226 Missense Mutation GAC,GGC D27G NP_003452.1
XM_011516569.2 226 Missense Mutation GAC,GGC D27G XP_011514871.2
XM_017012587.1 226 Missense Mutation GAC,GGC D86G XP_016868076.1

View Full Product Details