Product Details

SNP ID
rs117837628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144452205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCACTGCTCCTTAAATGCCACA[A/G]TCACGCGGGCCACTGCTCCTTTCAT
Phenotype
MIM: 606370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPK1 PubMed Links

Gene Details

Gene
TPK1
Gene Name
thiamin pyrophosphokinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042482.1 2404 UTR 3 NP_001035947.1
NM_022445.3 2404 UTR 3 NP_071890.2
XM_005249970.1 2404 UTR 3 XP_005250027.1
XM_011516031.1 2404 UTR 3 XP_011514333.1
XM_011516032.2 2404 UTR 3 XP_011514334.1
XM_011516033.2 2404 UTR 3 XP_011514335.1
XM_011516034.2 2404 UTR 3 XP_011514336.1
XM_011516035.2 2404 Intron XP_011514337.1
XM_011516037.2 2404 UTR 3 XP_011514339.1
XM_011516039.2 2404 UTR 3 XP_011514341.1
XM_011516040.2 2404 Intron XP_011514342.1
XM_011516041.2 2404 UTR 3 XP_011514343.1
XM_011516043.1 2404 UTR 3 XP_011514345.1
XM_011516044.1 2404 UTR 3 XP_011514346.1
XM_011516046.1 2404 Intron XP_011514348.1
XM_011516047.2 2404 UTR 3 XP_011514349.1
XM_011516048.1 2404 UTR 3 XP_011514350.1
XM_011516050.1 2404 UTR 3 XP_011514352.1
XM_017011969.1 2404 Intron XP_016867458.1
XM_017011970.1 2404 UTR 3 XP_016867459.1
XM_017011971.1 2404 UTR 3 XP_016867460.1
XM_017011972.1 2404 Intron XP_016867461.1
XM_017011973.1 2404 UTR 3 XP_016867462.1
XM_017011974.1 2404 UTR 3 XP_016867463.1
XM_017011975.1 2404 UTR 3 XP_016867464.1
XM_017011976.1 2404 UTR 3 XP_016867465.1
XM_017011977.1 2404 UTR 3 XP_016867466.1
XM_017011978.1 2404 UTR 3 XP_016867467.1
XM_017011979.1 2404 UTR 3 XP_016867468.1
XM_017011980.1 2404 UTR 3 XP_016867469.1
XM_017011981.1 2404 UTR 3 XP_016867470.1
XM_017011982.1 2404 UTR 3 XP_016867471.1
XM_017011983.1 2404 UTR 3 XP_016867472.1
XM_017011984.1 2404 UTR 3 XP_016867473.1
XM_017011985.1 2404 UTR 3 XP_016867474.1
XM_017011986.1 2404 UTR 3 XP_016867475.1

View Full Product Details