Product Details
- SNP ID
-
rs113056357
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:144453608 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTTTCCACAGTCACAACACCAGA[C/T]CCGTCGTAGGTATTGGAAGTACTGA
- Phenotype
-
MIM: 606370
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TPK1
PubMed Links
Gene Details
- Gene
- TPK1
- Gene Name
- thiamin pyrophosphokinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001042482.1 |
1001 |
Silent Mutation |
GGA,GGG |
G174G |
NP_001035947.1 |
NM_022445.3 |
1001 |
Silent Mutation |
GGA,GGG |
G223G |
NP_071890.2 |
XM_005249970.1 |
1001 |
Silent Mutation |
GGA,GGG |
G223G |
XP_005250027.1 |
XM_011516031.1 |
1001 |
Silent Mutation |
GGA,GGG |
G249G |
XP_011514333.1 |
XM_011516032.2 |
1001 |
Silent Mutation |
GGA,GGG |
G249G |
XP_011514334.1 |
XM_011516033.2 |
1001 |
Silent Mutation |
GGA,GGG |
G249G |
XP_011514335.1 |
XM_011516034.2 |
1001 |
Silent Mutation |
GGA,GGG |
G249G |
XP_011514336.1 |
XM_011516035.2 |
1001 |
UTR 3 |
|
|
XP_011514337.1 |
XM_011516037.2 |
1001 |
Silent Mutation |
GGA,GGG |
G244G |
XP_011514339.1 |
XM_011516039.2 |
1001 |
Silent Mutation |
GGA,GGG |
G223G |
XP_011514341.1 |
XM_011516040.2 |
1001 |
Intron |
|
|
XP_011514342.1 |
XM_011516041.2 |
1001 |
Silent Mutation |
GGA,GGG |
G218G |
XP_011514343.1 |
XM_011516043.1 |
1001 |
Silent Mutation |
GGA,GGG |
G200G |
XP_011514345.1 |
XM_011516044.1 |
1001 |
Silent Mutation |
GGA,GGG |
G169G |
XP_011514346.1 |
XM_011516046.1 |
1001 |
Intron |
|
|
XP_011514348.1 |
XM_011516047.2 |
1001 |
Silent Mutation |
GGA,GGG |
G117G |
XP_011514349.1 |
XM_011516048.1 |
1001 |
Silent Mutation |
GGA,GGG |
G117G |
XP_011514350.1 |
XM_011516050.1 |
1001 |
Silent Mutation |
GGA,GGG |
G117G |
XP_011514352.1 |
XM_017011969.1 |
1001 |
Intron |
|
|
XP_016867458.1 |
XM_017011970.1 |
1001 |
Silent Mutation |
GGA,GGG |
G244G |
XP_016867459.1 |
XM_017011971.1 |
1001 |
Silent Mutation |
GGA,GGG |
G244G |
XP_016867460.1 |
XM_017011972.1 |
1001 |
Intron |
|
|
XP_016867461.1 |
XM_017011973.1 |
1001 |
Silent Mutation |
GGA,GGG |
G223G |
XP_016867462.1 |
XM_017011974.1 |
1001 |
Silent Mutation |
GGA,GGG |
G223G |
XP_016867463.1 |
XM_017011975.1 |
1001 |
Silent Mutation |
GGA,GGG |
G223G |
XP_016867464.1 |
XM_017011976.1 |
1001 |
UTR 3 |
|
|
XP_016867465.1 |
XM_017011977.1 |
1001 |
Silent Mutation |
GGA,GGG |
G218G |
XP_016867466.1 |
XM_017011978.1 |
1001 |
Silent Mutation |
GGA,GGG |
G218G |
XP_016867467.1 |
XM_017011979.1 |
1001 |
Silent Mutation |
GGA,GGG |
G174G |
XP_016867468.1 |
XM_017011980.1 |
1001 |
Silent Mutation |
GGA,GGG |
G143G |
XP_016867469.1 |
XM_017011981.1 |
1001 |
Silent Mutation |
GGA,GGG |
G143G |
XP_016867470.1 |
XM_017011982.1 |
1001 |
Silent Mutation |
GGA,GGG |
G117G |
XP_016867471.1 |
XM_017011983.1 |
1001 |
Silent Mutation |
GGA,GGG |
G117G |
XP_016867472.1 |
XM_017011984.1 |
1001 |
Silent Mutation |
GGA,GGG |
G117G |
XP_016867473.1 |
XM_017011985.1 |
1001 |
Silent Mutation |
GGA,GGG |
G117G |
XP_016867474.1 |
XM_017011986.1 |
1001 |
Silent Mutation |
GGA,GGG |
G117G |
XP_016867475.1 |
View Full Product Details