Product Details

SNP ID
rs112563616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73570615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGCCCGCTTCAGGTGGCCCTG[A/C]ATCTCCTCCACCATGGCTCGGTGGC
Phenotype
MIM: 605842
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TBL2 PubMed Links

Gene Details

Gene
TBL2
Gene Name
transducin (beta)-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012453.2 1240 Missense Mutation ATG,ATT M412I NP_036585.1
XM_006715923.3 1240 Missense Mutation ATG,ATT M379I XP_006715986.1
XM_011516027.2 1240 Missense Mutation ATG,ATT M247I XP_011514329.1

View Full Product Details