Product Details

SNP ID
rs111453348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73686836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCAGCTGTGGCACTGGGCTGAG[C/T]GGAAGTTATCTGTCAGATGAAGGGC
Phenotype
MIM: 615733
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC30 PubMed Links

Gene Details

Gene
DNAJC30
Gene Name
DnaJ heat shock protein family (Hsp40) member C30
There are no transcripts associated with this gene.

Gene
WBSCR22
Gene Name
Williams-Beuren syndrome chromosome region 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202560.2 259 Silent Mutation AGC,AGT S67S NP_001189489.1
NM_017528.4 259 Silent Mutation AGC,AGT S67S NP_059998.2
XM_006715847.1 259 Silent Mutation AGC,AGT S90S XP_006715910.1
XM_011515778.1 259 Silent Mutation AGC,AGT S90S XP_011514080.1
XM_011515779.1 259 Silent Mutation AGC,AGT S67S XP_011514081.1

View Full Product Details