Product Details
- SNP ID
-
rs117964521
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:64214086 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAGCCCCAGAATATAAAGAGAAA[G/T]GAGATGGCAGCCAAACACCCAGGTA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF735
PubMed Links
Gene Details
- Gene
- ZNF735
- Gene Name
- zinc finger protein 735
View Full Product Details