Product Details

SNP ID
rs118145252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:88277559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCCTATTTGATGTGATATTATG[A/C]AGCCATTAAGAAAATATATTACGTA
Phenotype
MIM: 611098
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
STEAP4 PubMed Links

Gene Details

Gene
STEAP4
Gene Name
STEAP4 metalloreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205315.1 3442 UTR 3 NP_001192244.1
NM_001205316.1 3442 UTR 3 NP_001192245.1
NM_024636.3 3442 UTR 3 NP_078912.2

View Full Product Details