Product Details

SNP ID
rs112016765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93922387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGGCAGTGCCTGGGGACTGCC[A/G]GCTGATGTGGGGAAATCCTAGGAGG
Phenotype
MIM: 604390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNG11 PubMed Links

Gene Details

Gene
GNG11
Gene Name
G protein subunit gamma 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004126.3 Intron NP_004117.1

View Full Product Details