Product Details

SNP ID
rs115927539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99279652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCCTCACTGAGTGCCGCCAAGC[A/G]CAAGTTGGAAGGGGAGCTGAGCGAC
Phenotype
MIM: 608580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYH16 PubMed Links

Gene Details

Gene
MYH16
Gene Name
myosin heavy chain 16 pseudogene
There are no transcripts associated with this gene.

View Full Product Details