Product Details

SNP ID
rs113944890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100605089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCCGCTCCTCTCTCCCCTCCCC[A/T]GAGCACCAATTTTGCGGGGGGCGGC
Phenotype
MIM: 609097 MIM: 609125 MIM: 600270
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO24 PubMed Links

Gene Details

Gene
FBXO24
Gene Name
F-box protein 24
There are no transcripts associated with this gene.

Gene
MOSPD3
Gene Name
motile sperm domain containing 3
There are no transcripts associated with this gene.

Gene
PCOLCE
Gene Name
procollagen C-endopeptidase enhancer
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002593.3 Intron NP_002584.2
Gene
PCOLCE-AS1
Gene Name
PCOLCE antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details