Product Details

SNP ID
rs116815911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101195921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCTCAGTGCCTTGGGCTCAGGG[A/G]CTCAGCGAAAGGCCGCGGCCAGGCC
Phenotype
MIM: 610184
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOGAT3 PubMed Links

Gene Details

Gene
MOGAT3
Gene Name
monoacylglycerol O-acyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287147.1 1220 UTR 3 NP_001274076.1
NM_178176.3 1220 UTR 3 NP_835470.1
XM_005250309.3 1220 UTR 3 XP_005250366.1
XM_011516126.2 1220 UTR 3 XP_011514428.1
XM_011516127.2 1220 UTR 3 XP_011514429.1

View Full Product Details