Product Details

SNP ID
rs116714326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103082213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGTGAGAATATTCTAGCTTTGT[A/G]GCTGTGACATTTTCTTAACATTTCT
Phenotype
MIM: 611864 MIM: 609080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMC10 PubMed Links

Gene Details

Gene
ARMC10
Gene Name
armadillo repeat containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161009.2 Intron NP_001154481.1
NM_001161010.2 Intron NP_001154482.1
NM_001161011.2 Intron NP_001154483.1
NM_001161012.2 Intron NP_001154484.1
NM_001161013.2 Intron NP_001154485.1
NM_031905.4 Intron NP_114111.2
XM_005250628.2 Intron XP_005250685.1
XM_011516601.2 Intron XP_011514903.1
XM_011516602.2 Intron XP_011514904.1
XM_017012684.1 Intron XP_016868173.1
XM_017012685.1 Intron XP_016868174.1
XM_017012686.1 Intron XP_016868175.1
XM_017012687.1 Intron XP_016868176.1
XM_017012688.1 Intron XP_016868177.1
XM_017012689.1 Intron XP_016868178.1
Gene
FBXL13
Gene Name
F-box and leucine rich repeat protein 13
There are no transcripts associated with this gene.

View Full Product Details