Product Details

SNP ID
rs112874175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103472745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTCCTGATATCAGATGTAGTG[C/T]GAGATTTAAAAGAATGGAGAGCAAC
Phenotype
MIM: 600514
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927870 PubMed Links

Gene Details

Gene
LOC101927870
Gene Name
uncharacterized LOC101927870
There are no transcripts associated with this gene.

Gene
RELN
Gene Name
reelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005045.3 10610 UTR 3 NP_005036.2
NM_173054.2 10610 UTR 3 NP_774959.1

View Full Product Details