Product Details

SNP ID
rs112998546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:109959686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTGGCACTATACTGGTTGAGCT[C/T]TCCACTCTCATGGCCTGCAATGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF3IP1 PubMed Links

Gene Details

Gene
EIF3IP1
Gene Name
eukaryotic translation initiation factor 3 subunit I pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details