Product Details

SNP ID
rs117002623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112222592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTTTTCTTAGTTAGCTTAGC[C/T]AAGGGTTTGTCAATTTTGTGTATCT
Phenotype
MIM: 605465
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF277 PubMed Links

Gene Details

Gene
ZNF277
Gene Name
zinc finger protein 277
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021994.2 Intron NP_068834.2
XM_005250129.3 Intron XP_005250186.1
XM_011515768.2 Intron XP_011514070.1
XM_017011720.1 Intron XP_016867209.1

View Full Product Details