Product Details

SNP ID
rs113441026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:121351265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATGAGCCGCCGTGCCTCATCAT[C/T]GAGTCTTGAAGAGGGAGAGAAAGAA
Phenotype
MIM: 608618
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM3C PubMed Links

Gene Details

Gene
FAM3C
Gene Name
family with sequence similarity 3 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040020.1 812 Intron NP_001035109.1
NM_014888.2 812 Intron NP_055703.1
XM_011515736.2 812 Intron XP_011514038.1
XM_011515737.2 812 Missense Mutation AAT,GAT N158D XP_011514039.1

View Full Product Details