Product Details

SNP ID
rs113169335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128303817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAGGTCATTATTCTTCCCAGGC[A/G]GGGTCAACTTCCTCTATTAACACCC
Phenotype
MIM: 612074
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MGC27345 PubMed Links

Gene Details

Gene
MGC27345
Gene Name
uncharacterized protein MGC27345
There are no transcripts associated with this gene.

Gene
RBM28
Gene Name
RNA binding motif protein 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166135.1 Intron NP_001159607.1
NM_018077.2 Intron NP_060547.2
XM_011516370.2 Intron XP_011514672.1
XM_017012389.1 Intron XP_016867878.1
XM_017012390.1 Intron XP_016867879.1

View Full Product Details