Product Details

SNP ID
rs113686059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:129225928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGGTTTCCATACAGTTGATGA[C/T]TTTCTGCCTTTCTGTGGCCAACCGT
Phenotype
MIM: 616520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AHCYL2 PubMed Links

Gene Details

Gene
AHCYL2
Gene Name
adenosylhomocysteinase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130720.2 Intron NP_001124192.1
NM_001130722.2 Intron NP_001124194.2
NM_001130723.2 Intron NP_001124195.1
NM_015328.3 Intron NP_056143.1
XM_005250237.1 Intron XP_005250294.1
XM_011515987.2 Intron XP_011514289.1
XM_011515988.2 Intron XP_011514290.1
XM_017011904.1 Intron XP_016867393.1
XM_017011905.1 Intron XP_016867394.1
XM_017011906.1 Intron XP_016867395.1
XM_017011907.1 Intron XP_016867396.1

View Full Product Details