Product Details

SNP ID
rs112767101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:129439743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTCATGTAGATTAGATATGGA[C/T]ACAGACCATAGCTTTCATATATGTT
Phenotype
MIM: 616520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AHCYL2 PubMed Links

Gene Details

Gene
AHCYL2
Gene Name
adenosylhomocysteinase like 2
There are no transcripts associated with this gene.

Gene
STRIP2
Gene Name
striatin interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134336.1 Intron NP_001127808.1
NM_020704.2 Intron NP_065755.1
XM_006716069.3 Intron XP_006716132.1
XM_011516432.2 Intron XP_011514734.1
XM_011516435.1 Intron XP_011514737.1
XM_017012470.1 Intron XP_016867959.1
XM_017012471.1 Intron XP_016867960.1
XM_017012472.1 Intron XP_016867961.1

View Full Product Details